(S)-2-((2R,3R)-3-((S)-1-((3R,4S,5S)-4-((S)-N,3-Dimethyl-2-((S)-3-methyl-2-(methylamino)butanamido)butanamido)-3-methoxy-5-methylheptanoyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanamido)-3-phenylpropanoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Monomethyl Auristatin F
A309085-1MG
A309085-2MG
A309085-5MG
A309085-250MG
A309085-100MG
A309085-10MG
77015-128EA
18.04
USD
77015-128
77015-130
77015-132
77550-860
77683-911
77683-912
(S)-2-((2R,3R)-3-((S)-1-((3R,4S,5S)-4-((S)-N,3-Dimethyl-2-((S)-3-methyl-2-(methylamino)butanamido)butanamido)-3-methoxy-5-methylheptanoyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanamido)-3-phenylpropanoic acid 98%
(S)-2-((2R,3R)-3-((S)-1-((3R,4S,5S)-4-((S)-N,3-Dimethyl-2-((S)-3-methyl-2-(methylamino)butanamido)butanamido)-3-methoxy-5-methylheptanoyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanamido)-3-phenylpropanoic acid
Precaución: For research use only.
Formula:
C₃₉H₆₅N₅O₈ MW: 731.96 g/mol |
MDL Number:
MFCD25976742 CAS Number: 745017-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...