(2S,6S)-2,6-Dimethylmorpholine 97%

Supplier: Ambeed
Warning

A308986-1G A308986-250MG A308986-5G A308986-100MG
76988-918EA 167 USD
76988-918 76988-920 76988-922 77885-821
(2S,6S)-2,6-Dimethylmorpholine 97%
(2S,6S)-2,6-Dimethylmorpholine

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MDL Number: MFCD18250271
CAS Number: 276252-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR