4-(3,5-Dimethyl-1H-1,2,4-triazol-1-yl)benzoic acid 95%

Supplier: Ambeed
Warning

A308879-1G A308879-250MG A308879-5G
77015-122EA 375.56 USD
77015-122 77015-124 77015-126
4-(3,5-Dimethyl-1H-1,2,4-triazol-1-yl)benzoic acid 95%
4-(3,5-Dimethyl-1H-1,2,4-triazol-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁N₃O₂
MW: 217.22 g/mol
MDL Number: MFCD14630790
CAS Number: 858003-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR