6-Chloro-2-(trifluoromethyl)pyridin-3-amine 95%

Supplier: Ambeed
Danger

A308679-100MG A308679-5G A308679-1G
77015-112EA 62 USD
77015-112 77015-114 77890-592
6-Chloro-2-(trifluoromethyl)pyridin-3-amine 95%
6-Chloro-2-(trifluoromethyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MDL Number: MFCD18261523
CAS Number: 117519-13-8
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR