(S)-(-)-N-Boc-tert-leucinol 95%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl (1-hydroxy-3,3-dimethylbutan-2-yl)carbamate

A308596-1G A308596-5G A308596-250MG
77015-102EA 43.5 USD
77015-102 77015-104 77683-909
(S)-(-)-N-Boc-tert-leucinol 95%
(S)-(-)-N-Boc-tert-leucinol

Precaución: For research use only.

Formula: C₁₁H₂₃NO₃
MW: 217.309 g/mol
MDL Number: MFCD01861301
CAS Number: 153645-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR