1,2,4,5-Cyclohexanetetracarboxylic dianhydride 95%

Supplier: Ambeed
Warning

Synonyms: HPMDA, 1,2,4,5-Cyclohexanetetracarboxylic Dianhydride

A308528-100G A308528-1G A308528-25G A308528-500G A308528-5G
77002-656EA 198.39 USD
77002-656 77002-658 77002-660 77002-662 77002-664
1,2,4,5-Cyclohexanetetracarboxylic dianhydride 95%
1,2,4,5-Cyclohexanetetracarboxylic dianhydride

Precaución: for Research Use Only

Formula: C₁₀H₈O₆
MW: 224.17 g/mol
MDL Number: MFCD13704912
CAS Number: 2754-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR