(+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98%

Supplier: Ambeed
Warning

Synonyms: [3aR-[2(3′aR*,8′aS*),3′aβ,8′aβ]]-(+)-2,2′-Methylenebis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole], Bis((3aR,8aS)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)methane, (+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]

A308512-100MG A308512-1G A308512-250MG A308512-25G A308512-5G
76988-908EA 12.74 USD
76988-908 76988-910 76988-912 76988-914 76988-916
(+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98%
(+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]

Precaución: for Research Use Only

Formula: C₂₁H₁₈N₂O₂
MW: 330.39 g/mol
MDL Number: MFCD06797115
CAS Number: 180186-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR