3,4,5-Trifluorocinnamic acid 98%

Supplier: Ambeed
Warning

A308436-1G A308436-250MG A308436-25G
77002-650EA 14.86 USD
77002-650 77002-652 77002-654
3,4,5-Trifluorocinnamic acid 98%
3,4,5-Trifluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃O₂
MW: 202.13 g/mol
MDL Number: MFCD00083529
CAS Number: 152152-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR