(S)-2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride 95%

Supplier: Ambeed
Warning

A308140-100MG A308140-250MG A308140-1G
77015-098EA 105.03 USD
77015-098 77015-100 77683-908
(S)-2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride 95%
(S)-2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride

Precaución: For research use only.

Formula: C₇H₁₂Cl₂N₂O
MW: 211.08 g/mol
MDL Number: MFCD20487949
CAS Number: 1269652-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR