7-Chloro-1H-indole-2-carbonitrile 95%

Supplier: Ambeed
Warning

A308015-100MG A308015-250MG
77015-086EA 88.06 USD
77015-086 77015-088
7-Chloro-1H-indole-2-carbonitrile 95%
7-Chloro-1H-indole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅ClN₂
MW: 176.60 g/mol
CAS Number: 1264481-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR