2-Methyl-5-oxo-1-cyclopentenyl butyrate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-oxocyclopent-1-en-1-yl butyrate, Butyric Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester

A307817-10G A307817-25G A307817-5G
76988-894EA 47.75 USD
76988-894 76988-896 76988-898
2-Methyl-5-oxo-1-cyclopentenyl butyrate 98%
2-Methyl-5-oxo-1-cyclopentenyl butyrate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
MDL Number: MFCD00798052
CAS Number: 68227-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR