Methyl-3-bromo-4-hydroxybenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-bromo-4-hydroxybenzoate
A307790-100G
A307790-10G
A307790-1G
A307790-25G
A307790-500G
A307790-5G
77002-618EA
113.52
USD
77002-618
77002-620
77002-622
77002-624
77002-626
77002-628
Methyl-3-bromo-4-hydroxybenzoate 98%
Methyl-3-bromo-4-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₈H₇BrO₃ MW: 231.05 g/mol |
MDL Number:
MFCD06203850 CAS Number: 29415-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...