(4-(Aminomethyl)-3-fluorophenyl)boronic acid hydrochloride 97%

Supplier: Ambeed
Warning

A307341-1G A307341-250MG
77015-082EA 197.33 USD
77015-082 77015-084
(4-(Aminomethyl)-3-fluorophenyl)boronic acid hydrochloride 97%
(4-(Aminomethyl)-3-fluorophenyl)boronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀BClFNO₂
MW: 205.42 g/mol
MDL Number: MFCD11053810
CAS Number: 1072946-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR