4,4',4''-Trihydroxytriphenylmethane 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,4-Trihydroxytriphenylmethane, Tris(4-hydroxyphenyl)methane, 4,4',4''-Methylidynetrisphenol, Leucaurin

A306769-100G A306769-25G A306769-5G
77032-538EA 94.43 USD
77032-538 77032-540 77032-542
4,4',4''-Trihydroxytriphenylmethane 98%
4,4',4''-Trihydroxytriphenylmethane

Precaución: for Research Use Only

Formula: C₁₉H₁₆O₃
MW: 292.33 g/mol
MDL Number: MFCD00191589
CAS Number: 603-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR