2-(4-Oxo-4,5,6,7-tetrahydro-3H-cyclopenta[4,5]thieno[2,3-d]pyrimidin-3-yl)acetic acid 95%

Supplier: Ambeed
Warning

A306688-100MG A306688-250MG
77002-598EA 69 USD
77002-598 77002-600
2-(4-Oxo-4,5,6,7-tetrahydro-3H-cyclopenta[4,5]thieno[2,3-d]pyrimidin-3-yl)acetic acid 95%
2-(4-Oxo-4,5,6,7-tetrahydro-3H-cyclopenta[4,5]thieno[2,3-d]pyrimidin-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₃S
MW: 250.27 g/mol
MDL Number: MFCD02730276
CAS Number: 371206-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR