tert-Butyl 5-chloro-3-oxo-3H-spiro[isobenzofuran-1,4'-piperidine]-1'-carboxylate 95%

Supplier: Ambeed
Warning

A306612-100MG A306612-1G A306612-250MG
76988-882EA 44.56 USD
76988-882 76988-884 76988-886
tert-Butyl 5-chloro-3-oxo-3H-spiro[isobenzofuran-1,4'-piperidine]-1'-carboxylate 95%
tert-Butyl 5-chloro-3-oxo-3H-spiro[isobenzofuran-1,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₀ClNO₄
MW: 337.8 g/mol
MDL Number: MFCD24849714
CAS Number: 849106-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR