5-Chloro-1-methyl-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

A306308-100MG A306308-1G
76988-878EA 161.26 USD
76988-878 76988-880
5-Chloro-1-methyl-1H-pyrazol-3-amine 95%
5-Chloro-1-methyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₄H₆ClN₃
MW: 131.56 g/mol
MDL Number: MFCD19207885
CAS Number: 1191453-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR