5-Chloro-6-methoxypicolinic acid 95%

Supplier: Ambeed
Warning

A306222-100MG A306222-1G A306222-250MG A306222-5G
77032-526EA 25 USD
77032-526 77872-417 77872-418 77872-419
5-Chloro-6-methoxypicolinic acid 95%
5-Chloro-6-methoxypicolinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MDL Number: MFCD13185830
CAS Number: 1214373-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR