Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-hydroxy-2-methylthiazole-5-carboxylate, 4-Hydroxy-2-methylthiazole-5-carboxylic acid ethyl ester, Ethyl 2-methyl-4-oxo-4,5-dihydro-1,3-thiazole-5-carboxylate

A306079-250MG A306079-5G A306079-10G A306079-25G A306079-1G
77015-070EA 9.55 USD
77015-070 77015-072 77593-834 77593-836 77612-756
Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate 98%
Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₃S
MW: 187.216 g/mol
MDL Number: MFCD08275722
CAS Number: 20737-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR