2-Ethoxy-3-formyl-5-methylphenylboronic acid 95%

Supplier: Ambeed
Danger

Synonyms: (2-Ethoxy-3-formyl-5-methylphenyl)boronic acid

A305911-1G A305911-250MG A305911-5G
77015-060EA 65 USD
77015-060 77015-062 77015-064
2-Ethoxy-3-formyl-5-methylphenylboronic acid 95%
2-Ethoxy-3-formyl-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₄
MDL Number: MFCD08276749
CAS Number: 480424-54-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR