(S)-2-Aminopent-4-enoic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Allyl-Gly-OH·HCl, L-2-Allylglycine hydrochloride, H-Gly(Ally)-OH HCl, L-Allylglycine hydrocholride

A305508-1G A305508-250MG A305508-5G
77002-568EA 22 USD
77002-568 77002-570 77672-546
(S)-2-Aminopent-4-enoic acid hydrochloride 98%
(S)-2-Aminopent-4-enoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₂
MW: 151.59 g/mol
MDL Number: MFCD00002627
CAS Number: 195316-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR