6-(Trifluoromethoxy)isatin 97%

Supplier: Ambeed
Warning

A305438-100MG A305438-250MG A305438-1G
77015-040EA 8 USD
77015-040 77015-042 77890-584
6-(Trifluoromethoxy)isatin 97%
6-(Trifluoromethoxy)isatin

Precaución: for Research Use Only

Formula: C₉H₄F₃NO₃
MDL Number: MFCD06804530
CAS Number: 162252-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR