6-Bromo-2,3-dihydro-1H-inden-1-ol 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A305401-250MG A305401-5G A305401-1G A305401-25G
77015-036EA 21.22 USD
77015-036 77015-038 77593-830 77683-905
6-Bromo-2,3-dihydro-1H-inden-1-ol 98%, Ambeed.Inc
6-Bromo-2,3-dihydro-1H-inden-1-ol

Precaución: For research use only.

Formula: C₉H₉BrO
MW: 213.07 g/mol
MDL Number: MFCD02916451
CAS Number: 75476-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR