(S)-3-(Cbz-amino)piperidine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Benzyl piperidin-3-ylcarbamate

A305030-1G A305030-250MG A305030-5G
76988-848EA 50.93 USD
76988-848 76988-850 76988-852
(S)-3-(Cbz-amino)piperidine 97%
(S)-3-(Cbz-amino)piperidine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.3 g/mol
MDL Number: MFCD03426377
CAS Number: 478646-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR