Fexofenadine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)-2-methylpropanoic acid hydrochloride, Fexofenadine HCl

A304995-10MG A304995-1MG A304995-50MG
77032-520EA 8 USD
77032-520 77032-522 77032-524
Fexofenadine hydrochloride 98%
Fexofenadine hydrochloride

Precaución: for Research Use Only

Formula: C₃₂H₃₉NO₄
MDL Number: MFCD00865710
CAS Number: 153439-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR