(5-Methoxypyridin-2-yl)methanamine 95%

Supplier: Ambeed
Warning

A304613-100MG A304613-250MG
76988-844EA 97.61 USD
76988-844 76988-846
(5-Methoxypyridin-2-yl)methanamine 95%
(5-Methoxypyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.16 g/mol
MDL Number: MFCD10697534
CAS Number: 905306-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR