2-Cyano-4'-methylbiphenyl 98%

Supplier: Ambeed
Warning

A304604-1000G A304604-100G A304604-10G A304604-25G A304604-500G
77015-012EA 116 USD
77015-012 77015-014 77015-016 77015-018 77015-020
2-Cyano-4'-methylbiphenyl 98%
2-Cyano-4'-methylbiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MDL Number: MFCD00151805
CAS Number: 114772-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR