4-Bromothiophene-2-carboxamide 98%

Supplier: Ambeed
Warning

A304462-1G A304462-250MG A304462-5G
77032-508EA 28 USD
77032-508 77032-510 77032-512
4-Bromothiophene-2-carboxamide 98%
4-Bromothiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₄BrNOS
MDL Number: MFCD09859843
CAS Number: 83933-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR