Benzyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate 96%

Supplier: Ambeed
Warning

A303963-100MG A303963-1G A303963-250MG
76988-828EA 39 USD
76988-828 76988-830 76988-832
Benzyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate 96%
Benzyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₂₃BN₂O₄
MW: 354.21 g/mol
MDL Number: MFCD13195763
CAS Number: 1218790-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR