2-(Phenylamino)benzamide 97%

Supplier: Ambeed
Warning

Synonyms: SIRT-IN-3

A303109-1G A303109-250MG
77014-980EA 109.28 USD
77014-980 77014-982
2-(Phenylamino)benzamide 97%
2-(Phenylamino)benzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O
MW: 212.24 g/mol
MDL Number: MFCD00476529
CAS Number: 1211-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR