1-[(4-Nitrophenyl)sulfonyl]piperazine 98+%

Supplier: Ambeed
Warning

Synonyms: 1-((4-Nitrophenyl)sulphonyl)piperazine

A302944-100MG A302944-1G A302944-250MG
77014-974EA 21.22 USD
77014-974 77014-976 77014-978
1-[(4-Nitrophenyl)sulfonyl]piperazine 98+%
1-[(4-Nitrophenyl)sulfonyl]piperazine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃O₄S
MW: 271.3 g/mol
MDL Number: MFCD04059504
CAS Number: 403825-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR