tert-Butyl 4-(pyrimidin-4-yl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A302723-250MG A302723-1G
76988-810EA 24.41 USD
76988-810 77683-901
tert-Butyl 4-(pyrimidin-4-yl)piperazine-1-carboxylate 95%
tert-Butyl 4-(pyrimidin-4-yl)piperazine-1-carboxylate

Precaución: For research use only.

Formula: C₁₃H₂₀N₄O₂
MW: 264.32 g/mol
MDL Number: MFCD26383921
CAS Number: 221050-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR