Benzyl (5-hydroxypentyl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Z-Amino)-1-pentanol

A302500-100MG A302500-1G A302500-250MG A302500-5G
76988-800EA 31 USD
76988-800 76988-802 76988-804 77885-805
Benzyl (5-hydroxypentyl)carbamate 98%
Benzyl (5-hydroxypentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
MW: 237.29 g/mol
MDL Number: MFCD01863135
CAS Number: 87905-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR