5,6-Dimethoxyisobenzofuran-1(3H)-one 95%

Supplier: Ambeed
Warning

A302485-100MG A302485-10G A302485-1G A302485-250MG A302485-5G
76988-790EA 6 USD
76988-790 76988-792 76988-794 76988-796 76988-798
5,6-Dimethoxyisobenzofuran-1(3H)-one 95%
5,6-Dimethoxyisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD00086313
CAS Number: 531-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR