(2-Methylbenzo[d]oxazol-5-yl)methanol 95%

Supplier: Ambeed
Warning

A302357-100MG A302357-250MG
76988-782EA 61.54 USD
76988-782 77612-742
(2-Methylbenzo[d]oxazol-5-yl)methanol 95%
(2-Methylbenzo[d]oxazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.17 g/mol
MDL Number: MFCD12965126
CAS Number: 136663-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR