3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 95%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxycyclohexanecarboxylic acid, 3-Hydroxycyclohexane-1-carboxylic acid

A302294-1G A302294-250MG A302294-100MG
77014-968EA 108.22 USD
77014-968 77612-740 77683-900
3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 95%
3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture)

Precaución: For research use only.

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD19228828
CAS Number: 606488-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR