Fmoc-L-β-homoglutamic acid-d-t-butyl ester 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-β-HoGlu(OtBu)-OH, Fmoc-β-HomoGlu(OtBu)-OH
A302292-100MG
A302292-1G
A302292-250MG
A302292-5G
77002-542EA
28.65
USD
77002-542
77002-544
77002-546
77002-548
Fmoc-L-β-homoglutamic acid-d-t-butyl ester 98+%
Fmoc-L-beta-homoglutamic acid-d-t-butyl ester
Precaución: for Research Use Only
Formula:
C₂₅H₂₉NO₆ MW: 439.51 g/mol |
MDL Number:
MFCD01863053 CAS Number: 203854-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...