(R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A301811-1G A301811-250MG
76988-774EA 59 USD
76988-774 77890-574
(R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate hydrochloride 95%
(R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₁ClN₂O₄
MDL Number: MFCD11101236
CAS Number: 1251903-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR