6-(Trifluoromethyl)uracil 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)uracil, 2,4-Dihydroxy-6-trifluoromethylpyrimidine

A301694-1G A301694-5G A301694-10G
76988-770EA 9 USD
76988-770 76988-772 77593-820
6-(Trifluoromethyl)uracil 95%
6-(Trifluoromethyl)uracil

Precaución: for Research Use Only

Formula: C₅H₃F₃N₂O₂
MW: 180.09 g/mol
MDL Number: MFCD01011762
CAS Number: 672-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR