trans-Indole-3-acrylic acid 95%

Supplier: Ambeed
Warning

Synonyms: trans-3-Indoleacrylic acid

A301641-250MG A301641-25G A301641-5G A301641-1G
77002-526EA 16.98 USD
77002-526 77002-528 77002-530 77601-360
trans-Indole-3-acrylic acid 95%
trans-Indole-3-acrylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MW: 187.20 g/mol
MDL Number: MFCD00005633
CAS Number: 29953-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR