(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((2-phenylacetamido)methyl)thio)propanoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Cys(PhAcm)-OH
A301615-1G
A301615-250MG
A301615-5G
A301615-10G
76988-764EA
27
USD
76988-764
76988-766
76988-768
77885-800
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((2-phenylacetamido)methyl)thio)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((2-phenylacetamido)methyl)thio)propanoic acid
Precaución: for Research Use Only
Formula:
C₂₇H₂₆N₂O₅S MW: 490.57 g/mol |
MDL Number:
MFCD18427303 CAS Number: 159680-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...