4-(Trifluoromethyl)pyridine-2,6-diol 95%

Supplier: Ambeed
Warning

A301519-100MG A301519-1G A301519-250MG
77014-960EA 38.2 USD
77014-960 77014-962 77014-964
4-(Trifluoromethyl)pyridine-2,6-diol 95%
4-(Trifluoromethyl)pyridine-2,6-diol

Precaución: for Research Use Only

Formula: C₆H₄F₃NO₂
MW: 179.09 g/mol
MDL Number: MFCD11100755
CAS Number: 356518-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR