a-Tosyl(4-methoxybenzyl)isocyanide 95%

Supplier: Ambeed
Danger

Synonyms: a-Tosyl-(4-methoxybenzyl) isocyanide

A301512-250MG A301512-1G A301512-5G
77032-490EA 90 USD
77032-490 77872-405 77872-406
a-Tosyl(4-methoxybenzyl)isocyanide 95%
a-Tosyl(4-methoxybenzyl)isocyanide

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₃S
MDL Number: MFCD04114785
CAS Number: 263389-54-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR