Ethyl 4-ethoxy-2-oxo-1,2-dihydropyridine-3-carboxylate 98%

Supplier: Ambeed
Warning

A301156-1G A301156-250MG
77014-956EA 31 USD
77014-956 77014-958
Ethyl 4-ethoxy-2-oxo-1,2-dihydropyridine-3-carboxylate 98%
Ethyl 4-ethoxy-2-oxo-1,2-dihydropyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MW: 211.22 g/mol
MDL Number: MFCD18911742
CAS Number: 1174046-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR