Bis(benzonitrile)palladium(II) dichloride 99% Pd> 27.7%

Supplier: Ambeed
Danger

A301002-100MG A301002-1G A301002-250MG A301002-5G A301002-25G
77002-518EA 7 USD
77002-518 77002-520 77002-522 77002-524 77672-530
Bis(benzonitrile)palladium(II) dichloride 99% Pd> 27.7%
Bis(benzonitrile)dichloropalladium(II)

Precaución: for Research Use Only

Formula: C₁₄H₁₀Cl₂N₂Pd
MDL Number: MFCD00013123
CAS Number: 14220-64-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR