1-Boc-4-methylpiperidine 97%
Supplier: Ambeed
|
Danger
|
A300732-1G
A300732-250MG
A300732-5G
77014-946EA
32
USD
77014-946
77890-570
77890-571
1-Boc-4-methylpiperidine 97%
1-Boc-4-methylpiperidine
Precaución: for Research Use Only
Formula:
C₁₁H₂₁NO₂ |
MDL Number:
MFCD04114947 CAS Number: 123387-50-8 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...