6-Oxa-3-azabicyclo[3.1.1]heptane hydrochloride 97%

Supplier: Ambeed
Warning

A300634-100MG A300634-250MG A300634-50MG
77014-940EA 131.56 USD
77014-940 77014-942 77014-944
6-Oxa-3-azabicyclo[3.1.1]heptane hydrochloride 97%
6-Oxa-3-azabicyclo[3.1.1]heptane hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO
MW: 135.59 g/mol
MDL Number: MFCD22689277
CAS Number: 1414958-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR