4-Hydrazinotetrahydro-2H-pyran dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (Tetrahydro-2H-pyran-4-yl)hydrazine dihydrochloride, 4-Hydrazinooxane dihydrochloride

A300304-100MG A300304-1G A300304-250MG A300304-25G
77032-480EA 15.92 USD
77032-480 77032-482 77032-484 77032-486
4-Hydrazinotetrahydro-2H-pyran dihydrochloride 97%
4-Hydrazinotetrahydro-2H-pyran dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O·2ClH
MW: 189.083 g/mol
MDL Number: MFCD16621145
CAS Number: 1187974-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR