Methyl-2-aminothiazole-5-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-aminothiazole-5-carboxylate, 2-Aminothiazole-5-carboxylic acid methyll ester
A300179-100G
A300179-1G
A300179-25G
A300179-5G
A300179-10G
77014-926EA
155.96
USD
77014-926
77014-928
77014-930
77014-932
77683-895
Methyl-2-aminothiazole-5-carboxylate 95%
Methyl-2-aminothiazole-5-carboxylate
Precaución: For research use only.
Formula:
C₅H₆N₂O₂S MW: 158.1800 g/mol |
MDL Number:
MFCD03788562 CAS Number: 6633-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...