4',7-Dimethoxyisoflavone 97%

Supplier: Ambeed
Warning

Synonyms: 7-Methoxy-3-(4-methoxyphenyl)chromen-4-one, 7-Methoxy-3-(4-methoxyphenyl)-4-benzopyrone, 4',7-Dimethoxyisoflavone

A299621-100MG A299621-10MG A299621-1G A299621-1MG A299621-250MG A299621-5G A299621-5MG
77032-460EA 31.83 USD
77032-460 77032-462 77032-464 77032-466 77032-468 77032-470 77032-472
4',7-Dimethoxyisoflavone 97%
4',7-Dimethoxyisoflavone

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₄
MW: 282.3 g/mol
MDL Number: MFCD00075889
CAS Number: 1157-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR